Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptprf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019249.1 | NP_062122.1 | 31 | 6332 | ||
XM_006238677.2 | XP_006238739.1 | 35 | 7413 | |||
XM_006238678.2 | XP_006238740.1 | 34 | 7386 | |||
XM_006238680.2 | XP_006238742.1 | 33 | 7353 | |||
XM_006238681.2 | XP_006238743.1 | 32 | 7047 | |||
GenBank mRNA | L11586.1 | - | 31 | 6332 | ||
M60103.1 | - | 17 | 3215 | |||
U00477.1 | - | 10 | 2004 | |||
X83505.1 | - | 25 | 4494 |