Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other JUN SIRNA Assays ›
Gene Symbol:
Jun
Gene Name:
jun proto-oncogene
Gene Aliases:
-
Chromosome Location:
Chr.5: 114011184 - 114014277 on Build Rnor_6.0
UniGene ID:
Rn.93714
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200905
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Jun
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_021835.3
NP_068607.1
1
1382
GenBank mRNA
BC078738.1
-
1
619
7124370
X17163.1
-
1
876
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010591, AK159196, AK159220, BC021888, AK133867, and others.
Panther Classification:
Molecular Function -
basic leucine zipper transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
microglial cell activation
release of cytochrome c from mitochondria
liver development
positive regulation of endothelial cell proliferation
outflow tract morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transforming growth factor beta receptor signaling pathway
SMAD protein import into nucleus
aging
learning
circadian rhythm
positive regulation of cell proliferation
negative regulation of cell proliferation
response to radiation
response to mechanical stimulus
cellular process
response to organic substance
response to organic cyclic compound
monocyte differentiation
axon regeneration
negative regulation of protein autophosphorylation
response to lipopolysaccharide
response to cytokine
leading edge cell differentiation
response to muscle stretch
response to drug
response to hydrogen peroxide
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of DNA binding
negative regulation of neuron apoptotic process
positive regulation of neuron apoptotic process
negative regulation by host of viral transcription
positive regulation by host of viral transcription
positive regulation of monocyte differentiation
positive regulation of DNA replication
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fibroblast proliferation
positive regulation of smooth muscle cell proliferation
cellular response to potassium ion starvation
response to cAMP
regulation of cell cycle
membrane depolarization
SMAD protein signal transduction
cellular response to calcium ion
negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
DNA binding
chromatin binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription coactivator activity
protein binding
transcription factor binding
enzyme binding
cAMP response element binding
protein homodimerization activity
sequence-specific DNA binding
transcription regulatory region DNA binding
poly(A) RNA binding
protein heterodimerization activity
R-SMAD binding
HMG box domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0