Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012343.3 | NP_036475.3 | 22 | 3579 | ||
NM_182977.2 | NP_892022.2 | 22 | 3545 | |||
XM_005248274.4 | XP_005248331.1 | 22 | 3557 | |||
XM_005248275.4 | XP_005248332.1 | 17 | 2807 | |||
XM_006714461.3 | XP_006714524.1 | 21 | 3171 | |||
XM_011514001.2 | XP_011512303.1 | 22 | 3576 | |||
XM_011514002.2 | XP_011512304.1 | 21 | 3266 | |||
XM_017009293.1 | XP_016864782.1 | 22 | 3447 | |||
GenBank mRNA | AK297558.1 | - | 15 | 2192 | ||
AK299622.1 | - | 14 | 2009 | |||
AL831822.2 | - | 22 | 3546 | |||
BC032370.1 | - | 6 | 774 | |||
BC110543.1 | - | 22 | 3395 | 40036686 | ||
BC110544.1 | - | 21 | 3392 | 40036689 | ||
U40490.1 | - | 22 | 3466 | |||
Z50101.1 | - | 21 | 3367 |