Search
Search

Mrs2| Gene Symbol: Mrs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024001.1 | NP_076491.1 | 10 | 1190 | ||
| XM_006253937.2 | XP_006253999.1 | 10 | 1308 | |||
| GenBank mRNA | AF288289.1 | - | 10 | 1190 | ||
| AF288290.1 | - | 6 | 641 | |||
| AF288291.1 | - | 6 | 643 | |||
| BC083554.1 | - | 10 | 1183 | |||
| BC098916.1 | - | 10 | 1165 | |||