Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other JAK2 SIRNA Assays ›
Gene Symbol:
Jak2
Gene Name:
Janus kinase 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 247398667 - 247457521 on Build Rnor_6.0
UniGene ID:
Rn.18909
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
201101
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Jak2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031514.1
NP_113702.1
3
263
GenBank mRNA
U13396.1
-
3
263
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKK activity
activation of cysteine-type endopeptidase activity involved in apoptotic process
response to oxidative stress
signal transduction
enzyme linked receptor protein signaling pathway
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
JAK-STAT cascade
tyrosine phosphorylation of STAT protein
STAT protein import into nucleus
positive regulation of cell proliferation
negative regulation of cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
hormone-mediated signaling pathway
negative regulation of cardiac muscle cell apoptotic process
positive regulation of cell-substrate adhesion
positive regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
cytokine-mediated signaling pathway
negative regulation of cell-cell adhesion
myeloid cell differentiation
cell differentiation
erythrocyte differentiation
positive regulation of cell migration
axon regeneration
mineralocorticoid receptor signaling pathway
positive regulation of insulin secretion
response to lipopolysaccharide
positive regulation of phosphoprotein phosphatase activity
positive regulation of interleukin-1 beta production
positive regulation of tumor necrosis factor production
positive regulation of protein import into nucleus, translocation
response to hydroperoxide
tumor necrosis factor-mediated signaling pathway
host programmed cell death induced by symbiont
response to tumor necrosis factor
histone H3-Y41 phosphorylation
intracellular signal transduction
interleukin-12-mediated signaling pathway
peptidyl-tyrosine autophosphorylation
tyrosine phosphorylation of Stat3 protein
tyrosine phosphorylation of Stat5 protein
tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of tyrosine phosphorylation of Stat5 protein
activation of JAK2 kinase activity
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of DNA binding
negative regulation of DNA binding
negative regulation of neuron apoptotic process
positive regulation of nitric oxide biosynthetic process
positive regulation of cell differentiation
negative regulation of heart contraction
positive regulation of transcription from RNA polymerase II promoter
response to antibiotic
protein autophosphorylation
platelet-derived growth factor receptor signaling pathway
regulation of inflammatory response
positive regulation of inflammatory response
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of cell activation
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of nitric-oxide synthase biosynthetic process
interferon-gamma-mediated signaling pathway
growth hormone receptor signaling pathway
JAK-STAT cascade involved in growth hormone signaling pathway
positive regulation of growth hormone receptor signaling pathway
mammary gland epithelium development
response to interleukin-12
cellular response to lipopolysaccharide
cellular response to dexamethasone stimulus
extrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
positive regulation of apoptotic signaling pathway
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
growth hormone receptor binding
interleukin-12 receptor binding
protein binding
ATP binding
protein C-terminus binding
protein kinase binding
heme binding
type 1 angiotensin receptor binding
acetylcholine receptor binding
histone kinase activity (H3-Y41 specific)
SH2 domain binding
histone binding
phosphatidylinositol 3-kinase binding
insulin receptor substrate binding
peptide hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gjvvj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0