Search
Search

Gstm2| Gene Symbol: Gstm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177426.1 | NP_803175.1 | 358 | |||
| GenBank mRNA | FJ179402.1 | - | 358 | |||
| FQ214985.1 | - | 5, 6 | 391 | |||
| FQ218547.1 | - | 5, 6 | 424 | |||
| FQ218596.1 | - | 5, 6 | 391 | |||
| FQ219549.1 | - | 5, 6 | 393 | |||
| FQ219560.1 | - | 5, 6 | 391 | |||
| J02592.1 | - | 3, 4 | 262 | |||
| M13590.1 | - | 4, 5 | 288 | |||