Search
Search

| Gene Symbol: Gstm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177426.1 | NP_803175.1 | 380 | |||
| GenBank mRNA | BC158576.1 | - | 4 | 1589 | ||
| FJ179402.1 | - | 380 | ||||
| FQ214985.1 | - | 6 | 413 | |||
| FQ218547.1 | - | 6 | 446 | |||
| FQ218596.1 | - | 6 | 413 | |||
| FQ219549.1 | - | 6 | 415 | |||
| FQ219560.1 | - | 6 | 413 | |||
| J02592.1 | - | 4 | 284 | |||
| M13590.1 | - | 5 | 310 | |||
| Gene Symbol: Gstm6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106464.1 | NP_001099934.1 | 5 | 350 | ||