Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC2 SIRNA Assays ›
          Gene Symbol:
          HDAC2
          Gene Name:
          histone deacetylase 2
          Gene Aliases:
          HD2, RPD3, YAF1
          Chromosome Location:
          Chr.6: 113936156 - 113971195 on Build GRCh38
          UniGene ID:
          Hs.3352
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 213399

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HDAC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001527.3 NP_001518.3 13 1699
          NR_033441.1 14 1727
          NR_073443.1 13 1605
          XM_011535788.1 XP_011534090.1 14 1451
          XM_017010799.1 XP_016866288.1 14 1673
          GenBank mRNA AB209190.1 - 7 4415
          AK092156.1 - 13 1605
          AK097376.1 - 14 1661
          AK296856.1 - 13 1699
          BC031055.1 - 13 1663
          U31814.1 - 13 1616

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          response to amphetamine
          cardiac muscle hypertrophy
          chromatin remodeling
          maintenance of chromatin silencing
          transcription, DNA-templated
          blood coagulation
          positive regulation of cell proliferation
          epidermal cell differentiation
          positive regulation of epithelial to mesenchymal transition
          positive regulation of receptor biosynthetic process
          negative regulation of neuron projection development
          dendrite development
          histone deacetylation
          response to caffeine
          response to lipopolysaccharide
          positive regulation of interleukin-1 production
          positive regulation of tumor necrosis factor production
          circadian regulation of gene expression
          positive regulation of collagen biosynthetic process
          cellular response to heat
          response to nicotine
          response to cocaine
          odontogenesis of dentin-containing tooth
          response to drug
          positive regulation of tyrosine phosphorylation of Stat3 protein
          embryonic digit morphogenesis
          ATP-dependent chromatin remodeling
          negative regulation of apoptotic process
          negative regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of MHC class II biosynthetic process
          positive regulation of proteolysis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          behavioral response to ethanol
          positive regulation of oligodendrocyte differentiation
          response to hyperoxia
          hair follicle placode formation
          negative regulation of dendritic spine development
          eyelid development in camera-type eye
          fungiform papilla formation
          cellular response to hydrogen peroxide
          histone H3 deacetylation
          histone H4 deacetylation
          cellular response to retinoic acid
          cellular response to transforming growth factor beta stimulus
          regulation of signal transduction by p53 class mediator
          cellular response to dopamine
          negative regulation of peptidyl-lysine acetylation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          core promoter binding
          RNA polymerase II repressing transcription factor binding
          chromatin binding
          histone deacetylase activity
          protein binding
          transcription factor binding
          deacetylase activity
          enzyme binding
          heat shock protein binding
          nucleosomal DNA binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          protein deacetylase activity
          sequence-specific DNA binding
          poly(A) RNA binding
          NF-kappaB binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0