Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC25A47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207117.2 | NP_997000.2 | 6 | 1697 | ||
GenBank mRNA | AB073385.1 | - | 7 | 1621 | ||
AK092001.1 | - | 7 | 1607 | |||
AY569438.1 | - | 6 | 1697 | |||
BX248260.1 | - | 6 | 1686 |