Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPLP1 SIRNA Assays ›
          Gene Symbol:
          Rplp1
          Gene Name:
          ribosomal protein, large, P1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 66862143 - 66863476 on Build Rnor_6.0
          UniGene ID:
          Rn.973
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 217863

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rplp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007604.2 NP_001007605.1 1, 2 184
          GenBank mRNA BC058151.1 - 1, 2 187 6888011
          FQ209966.1 - 1, 2 186
          FQ210237.1 - 1, 2 183
          FQ210375.1 - 1, 2 183
          FQ210407.1 - 1, 2 183
          FQ210632.1 - 1, 2 185
          FQ210864.1 - 1, 2 183
          FQ210988.1 - 1, 2 186
          FQ211327.1 - 1, 2 184
          FQ211648.1 - 1, 2 183
          FQ212351.1 - 1, 2 183
          FQ214973.1 - 1, 2 186
          FQ215065.1 - 1, 2 183
          FQ216905.1 - 1, 2 187
          FQ216921.1 - 1, 2 186
          FQ216959.1 - 1, 2 182
          FQ216979.1 - 1, 2 186
          FQ217216.1 - 1, 2 186
          FQ217313.1 - 1, 2 184
          FQ220167.1 - 1, 2 184
          FQ220212.1 - 1, 2 185
          FQ220998.1 - 1, 2 183
          FQ221027.1 - 1, 2 184
          FQ221120.1 - 1, 2 184
          FQ221137.1 - 1, 2 183
          FQ221201.1 - 1, 2 186
          FQ221359.1 - 1, 2 184
          FQ221455.1 - 1, 2 188
          FQ221534.1 - 1, 2 181
          FQ221567.1 - 1, 2 186
          FQ221595.1 - 1, 2 184
          FQ221620.1 - 1, 2 183
          FQ221698.1 - 1, 2 190
          FQ221717.1 - 1, 2 183
          FQ221756.1 - 1, 2 183
          FQ221784.1 - 1, 2 183
          FQ221894.1 - 1, 2 183
          FQ221969.1 - 1, 2 183
          FQ221980.1 - 1, 2 183
          FQ222031.1 - 1, 2 183
          FQ222178.1 - 1, 2 183
          FQ222307.1 - 1, 2 185
          FQ222468.1 - 1, 2 185
          FQ222559.1 - 1, 2 183
          FQ222608.1 - 1, 2 186
          FQ222618.1 - 1, 2 186
          FQ222784.1 - 1, 2 183
          FQ222801.1 - 1, 2 184
          FQ222805.1 - 1, 2 183
          FQ222816.1 - 1, 2 184
          FQ222879.1 - 1, 2 179
          FQ222923.1 - 1, 2 184
          FQ222965.1 - 1, 2 183
          FQ222966.1 - 1, 2 182
          FQ223013.1 - 1, 2 184
          FQ223066.1 - 1, 2 185
          FQ223337.1 - 1, 2 183
          FQ223344.1 - 1, 2 184
          FQ223388.1 - 1, 2 183
          FQ223390.1 - 1, 2 184
          FQ223470.1 - 1, 2 186
          FQ223512.1 - 1, 2 184
          FQ223537.1 - 1, 2 184
          FQ223567.1 - 1, 2 183
          FQ223591.1 - 1, 2 183
          FQ223674.1 - 1, 2 165
          FQ223769.1 - 1, 2 184
          FQ224482.1 - 1, 2 153
          FQ224625.1 - 1, 2 186
          FQ224766.1 - 1, 2 184
          FQ228323.1 - 1, 2 183
          FQ228361.1 - 1, 2 183
          FQ228390.1 - 1, 2 184
          FQ228490.1 - 1, 2 184
          FQ228523.1 - 1, 2 183
          FQ228528.1 - 1, 2 180
          FQ228539.1 - 1, 2 183
          FQ228605.1 - 1, 2 187
          FQ228650.1 - 1, 2 183
          FQ228792.1 - 1, 2 186
          FQ228990.1 - 1, 2 190
          FQ229009.1 - 1, 2 183
          FQ229050.1 - 1, 2 183
          FQ229318.1 - 1, 2 184
          FQ230089.1 - 1, 2 184
          X15097.1 - 1, 2 154

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_018853, BC058685, AK010656, BC092088, AK007832, and others.

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          structural constituent of ribosome

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0