Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vom1r19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008954.1 | NP_001008954.1 | 1 | 667 | ||
GenBank mRNA | AY510367.1 | - | 1 | 667 | ||
AY510368.1 | - | 1 | 667 |