Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zc3h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198631.2 | NP_941033.2 | 14 | 3596 | ||
GenBank mRNA | AK129275.1 | - | 8 | 2471 | ||
AK136235.1 | - | 1 | 1054 | |||
BC057627.1 | - | 5 | 1197 | 6412890 | ||
BC060274.1 | - | 2 | 1515 |