Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TPTE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290224.1 | NP_001277153.1 | 16 | 1443 | ||
NM_199259.3 | NP_954868.1 | 20 | 1665 | |||
NM_199260.3 | NP_954869.1 | 19 | 1605 | |||
NM_199261.3 | NP_954870.2 | 21 | 1719 | |||
GenBank mRNA | AF007118.1 | - | 21 | 1667 | ||
AF495908.1 | - | 20 | 1613 | |||
AK308254.1 | - | 20 | 1562 | |||
AK314285.1 | - | 21 | 1618 | |||
AL833345.1 | - | 21 | 1626 | |||
AY219886.1 | - | 20 | 1613 | |||
AY219887.1 | - | 19 | 1553 | |||
AY219888.1 | - | 16 | 1391 | |||
BC028719.1 | - | 20 | 1648 | 5269598 |