Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NGRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033088.1 | NP_001028260.2 | 3 | 346 | ||
NR_028052.1 | 2 | 757 | ||||
GenBank mRNA | AB029315.1 | - | 2 | 755 | ||
AF225423.1 | - | 2 | 646 | |||
AF242770.1 | - | 2 | 711 | |||
AK312641.1 | - | 3 | 560 | |||
AL360142.1 | - | 2 | 729 | |||
AL834503.1 | - | 2 | 746 | |||
AY049780.1 | - | 2 | 80 | |||
BC001682.2 | - | 3 | 328 | 2964457 | ||
BC007222.1 | - | 2 | 732 | 2819984 | ||
BC009389.2 | - | 2 | 707 | 4130266 | ||
BC017192.2 | - | 2 | 712 | 3139180 | ||
BC019023.2 | - | 2 | 713 | |||
BC096824.1 | - | 3 | 263 | |||
CD242640.1 | - | 3 | 333 |