Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RALGAPA1 SIRNA Assays ›
Gene Symbol:
RALGAPA1
Gene Name:
Ral GTPase activating protein catalytic alpha subunit 1
Gene Aliases:
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
Chromosome Location:
Chr.14: 35538352 - 35809304 on Build GRCh38
UniGene ID:
Hs.113150
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
251680
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RALGAPA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283043.1
NP_001269972.1
19
2974
NM_001283044.1
NP_001269973.1
19
3076
NM_014990.1
NP_055805.1
18
2935
NM_194301.2
NP_919277.2
18
2935
XM_005267491.3
XP_005267548.1
19
3154
XM_005267492.3
XP_005267549.1
19
3154
XM_005267493.3
XP_005267550.1
18
3013
XM_005267498.4
XP_005267555.1
4
764
XM_006720098.2
XP_006720161.1
19
4531
XM_006720099.2
XP_006720162.1
19
4531
XM_006720100.3
XP_006720163.1
18
4390
XM_006720101.2
XP_006720164.1
19
4531
XM_006720102.3
XP_006720165.1
19
4237
XM_006720103.2
XP_006720166.1
19
3154
XM_006720104.3
XP_006720167.1
19
4531
XM_011536615.2
XP_011534917.1
19
4531
XM_011536616.2
XP_011534918.1
19
4531
XM_017021141.1
XP_016876630.1
18
4390
XM_017021142.1
XP_016876631.1
18
4390
XM_017021143.1
XP_016876632.1
19
4531
XM_017021144.1
XP_016876633.1
19
3154
XM_017021145.1
XP_016876634.1
18
3013
XM_017021146.1
XP_016876635.1
4
2200
XM_017021147.1
XP_016876636.1
4
1086
GenBank mRNA
AB020691.1
-
14
2204
AB511280.1
-
19
2685
AY596970.1
-
18
2935
AY596971.1
-
18
2935
BC150596.1
-
19
2682
9056966
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
regulation of small GTPase mediated signal transduction
activation of GTPase activity
GTPase activator activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0