Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Herpud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024988.1 | NP_001020159.1 | 8 | 1499 | ||
XM_006242723.2 | XP_006242785.1 | 8 | 1680 | |||
XM_006242724.2 | XP_006242786.1 | 8 | 1742 | |||
GenBank mRNA | BC081861.1 | - | 8 | 1499 | 7134757 |