Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SHROOM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020717.3 | NP_065768.2 | 4 | 2444 | ||
NR_027121.1 | 4 | 2444 | ||||
XM_017029682.1 | XP_016885171.1 | 4 | 2550 | |||
XM_017029683.1 | XP_016885172.1 | 3 | 2415 | |||
XM_017029684.1 | XP_016885173.1 | 3 | 2567 | |||
XM_017029685.1 | XP_016885174.1 | 4 | 2567 | |||
XM_017029686.1 | XP_016885175.1 | 2 | 2432 | |||
XM_017029687.1 | XP_016885176.1 | 4 | 2554 | |||
GenBank mRNA | AB033028.1 | - | 4 | 2444 | ||
AY044234.1 | - | 3 | 2525 | |||
BC151240.1 | - | 4 | 2455 | 8860513 |