Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019063.4 | NP_061936.2 | 4 | 652 | ||
XM_005264267.1 | XP_005264324.1 | 4 | 632 | |||
XM_005264268.3 | XP_005264325.1 | 4 | 632 | |||
XM_006711992.2 | XP_006712055.1 | 5 | 753 | |||
GenBank mRNA | AF177377.2 | - | 4 | 614 | ||
AK314702.1 | - | 4 | 626 | |||
BC104647.1 | - | 4 | 579 | |||
BC140845.1 | - | 4 | 575 | 9021713 |