Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPNT SIRNA Assays ›
          Gene Symbol:
          NPNT
          Gene Name:
          nephronectin
          Gene Aliases:
          EGFL6L, POEM
          Chromosome Location:
          Chr.4: 105895440 - 105971671 on Build GRCh38
          UniGene ID:
          Hs.518921
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 279368

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPNT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001033047.2 NP_001028219.1 12 2537
          NM_001184690.1 NP_001171619.1 13 2588
          NM_001184691.1 NP_001171620.1 13 2627
          NM_001184692.1 NP_001171621.1 11 2450
          NM_001184693.1 NP_001171622.1 12 2540
          XM_005262888.4 XP_005262945.1 14 2665
          XM_005262890.1 XP_005262947.1 12 2312
          XM_011531820.2 XP_011530122.1 14 2668
          XM_011531822.2 XP_011530124.1 13 2617
          XM_011531823.2 XP_011530125.1 13 2581
          XM_011531824.2 XP_011530126.1 13 2578
          XM_011531825.2 XP_011530127.1 12 2527
          XM_017007984.1 XP_016863473.1 13 2411
          GenBank mRNA AK172856.1 - 1 273
          AL832465.1 - 12 2450

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ureteric bud development
          branching involved in ureteric bud morphogenesis
          cell-matrix adhesion
          positive regulation of alkaline phosphatase activity
          positive regulation of cell-substrate adhesion
          extracellular matrix organization
          positive regulation of transforming growth factor beta receptor signaling pathway
          cell-cell adhesion mediated by integrin
          establishment of protein localization
          positive regulation of osteoblast differentiation
          positive regulation of smooth muscle contraction
          positive regulation of ERK1 and ERK2 cascade
          cellular response to tumor necrosis factor
          pilomotor reflex
          positive regulation of transcription from RNA polymerase II promoter involved in smooth muscle cell differentiation
          integrin binding
          calcium ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0