Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RGS7BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029875.2 | NP_001025046.1 | 6 | 2402 | ||
NM_001271890.1 | NP_001258819.1 | 4 | 1516 | |||
NM_001271891.1 | NP_001258820.1 | 4 | 1514 | |||
NR_073511.1 | 5 | 1601 | ||||
GenBank mRNA | AK294808.1 | - | 5 | 1507 | ||
AK296494.1 | - | 4 | 1484 | |||
BC140707.1 | - | 6 | 1749 | 9021575 | ||
BX640900.1 | - | 6 | 1640 |