Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DST SIRNA Assays ›
Gene Symbol:
DST
Gene Name:
dystonin
Gene Aliases:
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
Chromosome Location:
Chr.6: 56457987 - 56954671 on Build GRCh38
UniGene ID:
Hs.604915
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28477
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144769.2
NP_001138241.1
28
3766
NM_001144770.1
NP_001138242.1
26
3437
NM_001723.5
NP_001714.1
15
2321
NM_015548.4
NP_056363.2
15
2321
NM_183380.3
NP_899236.1
25
3711
XM_005249310.3
XP_005249367.1
29
3860
XM_005249315.3
XP_005249372.1
28
3807
XM_005249316.2
XP_005249373.1
21
4083
XM_005249318.2
XP_005249375.1
27
3521
XM_005249319.2
XP_005249376.1
26
3422
XM_005249320.4
XP_005249377.1
26
3478
XM_005249322.4
XP_005249379.1
26
3373
XM_005249323.3
XP_005249380.1
25
3706
XM_005249324.2
XP_005249381.1
15
2318
XM_011514824.2
XP_011513126.1
28
3771
XM_011514825.2
XP_011513127.1
28
3767
XM_011514826.2
XP_011513128.1
25
3706
XM_017011205.1
XP_016866694.1
29
3870
XM_017011206.1
XP_016866695.1
29
3870
XM_017011207.1
XP_016866696.1
29
3807
XM_017011208.1
XP_016866697.1
29
3870
XM_017011209.1
XP_016866698.1
29
3870
XM_017011210.1
XP_016866699.1
29
3870
XM_017011211.1
XP_016866700.1
29
3870
XM_017011212.1
XP_016866701.1
29
3870
XM_017011213.1
XP_016866702.1
29
3870
XM_017011214.1
XP_016866703.1
29
3870
XM_017011215.1
XP_016866704.1
29
3870
XM_017011216.1
XP_016866705.1
29
3870
XM_017011217.1
XP_016866706.1
26
3451
XM_017011219.1
XP_016866708.1
29
3866
XM_017011220.1
XP_016866709.1
28
3809
XM_017011221.1
XP_016866710.1
29
3866
XM_017011222.1
XP_016866711.1
21
4025
XM_017011223.1
XP_016866712.1
29
3866
XM_017011224.1
XP_016866713.1
25
3706
XM_017011225.1
XP_016866714.1
29
3880
GenBank mRNA
AF400226.1
-
15
2213
AF400227.1
-
15
2213
AK299898.1
-
26
3437
AU117550.1
-
5
584
BC065536.1
-
15
2285
L11690.1
-
15
2315
M63618.1
-
5
552
M69225.1
-
15
2251
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
cell adhesion
integrin-mediated signaling pathway
response to wounding
maintenance of cell polarity
intermediate filament cytoskeleton organization
cell motility
actin binding
integrin binding
calcium ion binding
protein binding
protein C-terminus binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0