Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LCN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001712.2 | NP_001001712.2 | 5 | 558 | ||
GenBank mRNA | AK091788.1 | - | 2 | 1046 | ||
AL832485.1 | - | 1 | 799 | |||
AY301271.1 | - | 4 | 463 | |||
BC036875.1 | - | 2 | 160 | |||
BC040963.1 | - | 5 | 1803 | |||
BC046377.1 | - | 1 | 88 | |||
BC133045.1 | - | 5 | 558 | 40147067 | ||
BC144440.1 | - | 3 | 340 | |||
BC144441.1 | - | 4 | 450 | 9052964 | ||
BC171741.1 | - | 3 | 278 |