Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNF4A SIRNA Assays ›
          Gene Symbol:
          HNF4A
          Gene Name:
          hepatocyte nuclear factor 4 alpha
          Gene Aliases:
          FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14
          Chromosome Location:
          Chr.20: 44355801 - 44432845 on Build GRCh38
          UniGene ID:
          Hs.116462
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 290203

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HNF4A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000457.4 NP_000448.3 7 959
          NM_001030003.2 NP_001025174.1 7 780
          NM_001030004.2 NP_001025175.1 7 780
          NM_001258355.1 NP_001245284.1 8 1050
          NM_001287182.1 NP_001274111.1 8 1002
          NM_001287183.1 NP_001274112.1 8 1002
          NM_001287184.1 NP_001274113.1 8 1002
          NM_175914.4 NP_787110.2 7 780
          NM_178849.2 NP_849180.1 7 959
          NM_178850.2 NP_849181.1 7 959
          XM_005260407.3 XP_005260464.1 8 1079
          GenBank mRNA AB307703.1 - 7 815
          AY680696.1 - 7 780
          AY680697.1 - 7 780
          AY680698.1 - 7 780
          BC137539.1 - 7 881 9021539
          BC137540.1 - 7 881 9021540
          HQ692860.1 - 7 842
          HQ692868.1 - 7 776
          HQ692869.1 - 7 842
          X76930.1 - 7 834
          X87870.1 - 7 914
          X87871.1 - 7 899
          X87872.1 - 7 935
          Z49825.1 - 8 1035

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          ornithine metabolic process
          lipid metabolic process
          acyl-CoA metabolic process
          xenobiotic metabolic process
          establishment of tissue polarity
          sex differentiation
          blood coagulation
          negative regulation of cell proliferation
          response to glucose
          regulation of gastrulation
          regulation of lipid metabolic process
          signal transduction involved in regulation of gene expression
          cell differentiation
          negative regulation of cell growth
          intracellular receptor signaling pathway
          regulation of microvillus assembly
          negative regulation of protein import into nucleus, translocation
          response to drug
          negative regulation of tyrosine phosphorylation of Stat5 protein
          glucose homeostasis
          steroid hormone mediated signaling pathway
          negative regulation of sequence-specific DNA binding transcription factor activity
          cell-cell junction organization
          positive regulation of gluconeogenesis
          positive regulation of fatty acid biosynthetic process
          positive regulation of transcription, DNA-templated
          negative regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          regulation of insulin secretion
          response to cAMP
          lipid homeostasis
          phospholipid homeostasis
          SMAD protein signal transduction
          triglyceride homeostasis
          negative regulation of activation of JAK2 kinase activity
          positive regulation of cholesterol homeostasis
          fatty-acyl-CoA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II activating transcription factor binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          receptor binding
          fatty acid binding
          protein binding
          zinc ion binding
          palmitoyl-CoA hydrolase activity
          protein homodimerization activity
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0