Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTBP2 SIRNA Assays ›
          Gene Symbol:
          PTBP2
          Gene Name:
          polypyrimidine tract binding protein 2
          Gene Aliases:
          PTBLP, brPTB, nPTB
          Chromosome Location:
          Chr.1: 96721605 - 96815049 on Build GRCh38
          UniGene ID:
          Hs.596061
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 29271

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTBP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001300985.1 NP_001287914.1 12 1559
          NM_001300986.1 NP_001287915.1 12 1568
          NM_001300987.1 NP_001287916.1 13 1715
          NM_001300988.1 NP_001287917.1 12 1559
          NM_001300989.1 NP_001287918.1 12 1544
          NM_001300990.1 NP_001287919.1 11 1468
          NM_021190.3 NP_067013.1 12 1544
          NR_125356.1 11 1402
          NR_125357.1 11 1510
          XM_011541874.2 XP_011540176.1 12 1392
          XM_011541875.2 XP_011540177.1 12 1390
          XM_011541876.2 XP_011540178.1 12 1422
          XM_017001951.1 XP_016857440.1 11 1422
          GenBank mRNA AB051232.1 - 12 1366
          AB051233.1 - 11 1276
          AF176085.1 - 12 1336
          AF530580.1 - 12 1354
          AF530581.1 - 12 1354
          AF530582.1 - 4 407
          AF530583.1 - 4 422
          AF530584.1 - 4 407
          AF530585.1 - 4 422
          AK022716.1 - 7 850
          AK025333.1 - 2 207
          AK092216.1 - 11 1408
          AK295384.1 - 11 1323
          AK299755.1 - 11 1153
          AK299897.1 - 13 1496
          AK299922.1 - 12 1382
          BC016582.2 - 12 1313 3872881

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001005555, XM_006233238, XM_006233236, XM_006233237, AJ010585, and others.
          This siRNA also targets the following transcripts in Mouse: NM_019550, AF095718, BC010255, AK013734, AK137348, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splice site selection
          spinal cord development
          cerebellum development
          negative regulation of RNA splicing
          regulation of neural precursor cell proliferation
          nucleotide binding
          mRNA binding
          pre-mRNA binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lvxnv:80/100.66.77.23:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0