Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009877.2 | NP_001009877.2 | 5, 6 | 608 | ||
NM_001317951.1 | NP_001304880.1 | 5, 6 | 900 | |||
NM_023924.4 | NP_076413.3 | 5, 6 | 781 | |||
NR_134296.1 | 5, 6 | 1117 | ||||
XM_006714494.3 | XP_006714557.1 | 5, 6 | 760 | |||
XM_017009780.1 | XP_016865269.1 | 5, 6 | 760 | |||
GenBank mRNA | AK023503.1 | - | 5, 6 | 878 | ||
AK024392.1 | - | 2, 3 | 616 | |||
AK297573.1 | - | 5, 6 | 595 | |||
AK299157.1 | - | 5, 6 | 781 | |||
AK301987.1 | - | 5, 6 | 732 | |||
BC041590.1 | - | 3, 4 | 467 | 5428011 | ||
BX648703.1 | - | 5, 6 | 1119 | |||
DQ248311.1 | - | 1, 2 | 106 |