Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZC3H12A SIRNA Assays ›
          Gene Symbol:
          ZC3H12A
          Gene Name:
          zinc finger CCCH-type containing 12A
          Gene Aliases:
          MCPIP, MCPIP1, dJ423B22.1
          Chromosome Location:
          Chr.1: 37474518 - 37484377 on Build GRCh38
          UniGene ID:
          Hs.656294
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 32160

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZC3H12A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001323550.1 NP_001310479.1 2 308
          NM_001323551.1 NP_001310480.1 2 311
          NM_025079.2 NP_079355.2 2 311
          XM_011542198.2 XP_011540500.1 2 311
          GenBank mRNA AK026884.1 - 2 277
          AY920403.1 - 1 161
          BC005001.1 - 2 235 2958199
          CR457364.1 - 1 161
          FJ695517.1 - 2 249

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          endoribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, endonucleolytic cleavage-dependent decay
          angiogenesis
          negative regulation of protein phosphorylation
          positive regulation of defense response to virus by host
          transcription, DNA-templated
          apoptotic process
          inflammatory response
          cellular response to DNA damage stimulus
          nervous system development
          regulation of gene expression
          positive regulation of autophagy
          positive regulation of endothelial cell migration
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of muscle cell apoptotic process
          positive regulation of lipid storage
          positive regulation of cell death
          protein deubiquitination
          cell differentiation
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          cellular response to oxidative stress
          cellular response to glucose starvation
          positive regulation of protein import into nucleus
          negative regulation of NF-kappaB import into nucleus
          negative regulation of macrophage activation
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation by host of viral genome replication
          negative regulation of nitric oxide biosynthetic process
          positive regulation of fat cell differentiation
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of interleukin-1 beta secretion
          protein oligomerization
          defense response to virus
          negative regulation of cardiac muscle contraction
          positive regulation of mRNA catabolic process
          3'-UTR-mediated mRNA destabilization
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          RNA phosphodiester bond hydrolysis
          RNA phosphodiester bond hydrolysis, endonucleolytic
          RNA phosphodiester bond hydrolysis, exonucleolytic
          cellular response to virus
          negative regulation of cytokine production involved in inflammatory response
          positive regulation of execution phase of apoptosis
          negative regulation of interleukin-6 secretion
          positive regulation of p38MAPK cascade
          negative regulation of interferon-gamma secretion
          positive regulation of protein deubiquitination
          negative regulation of production of miRNAs involved in gene silencing by miRNA
          cellular response to sodium arsenite
          negative regulation of tumor necrosis factor secretion
          cellular response to phorbol 13-acetate 12-myristate
          cellular response to ionomycin
          cellular response to chemokine
          positive regulation of reactive oxygen species metabolic process
          positive regulation of miRNA catabolic process
          DNA binding
          chromatin binding
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          endoribonuclease activity
          exoribonuclease activity
          ribonuclease activity
          thiol-dependent ubiquitin-specific protease activity
          protein binding
          miRNA binding
          RNA stem-loop binding
          mRNA 3'-UTR AU-rich region binding
          ribosome binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0