Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FCRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159488.1 | NP_001152960.1 | 6, 7 | 1229 | ||
NM_030764.3 | NP_110391.2 | 6, 7 | 1229 | |||
XM_011509974.2 | XP_011508276.1 | 6, 7 | 1564 | |||
XM_011509975.2 | XP_011508277.1 | 6, 7 | 1566 | |||
XM_011509976.2 | XP_011508278.1 | 6, 7 | 1565 | |||
XM_017002316.1 | XP_016857805.1 | 6, 7 | 1564 | |||
XM_017002317.1 | XP_016857806.1 | 6, 7 | 1367 | |||
XM_017002319.1 | XP_016857808.1 | 5, 6 | 1279 | |||
GenBank mRNA | AF319438.1 | - | 1, 2 | 714 | ||
AF319439.1 | - | 1, 2 | 552 | |||
AF319440.1 | - | 1 | 961 | |||
AF390037.1 | - | 1, 2 | 552 | |||
AF459633.1 | - | 6, 7 | 1217 | |||
AK301113.1 | - | 6, 7 | 1216 | |||
AL833194.1 | - | 1, 2 | 3088 | |||
AY043465.1 | - | 6, 7 | 1216 | |||
AY358130.1 | - | 6, 7 | 1219 | |||
BC129836.1 | - | 1, 2 | 554 | 40115446 |