Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032125.2 | NP_115501.2 | 6 | 832 | ||
NR_037576.1 | 7 | 985 | ||||
NR_037577.1 | 7 | 920 | ||||
NR_037580.1 | 8 | 1073 | ||||
GenBank mRNA | AF251699.1 | - | 6 | 727 | ||
AK095889.1 | - | 2 | 2061 | |||
AK123218.1 | - | 3 | 2080 | |||
AK222642.1 | - | 6 | 711 | |||
AK303821.1 | - | 6 | 786 | |||
AK307077.1 | - | 7 | 823 | |||
AL136683.1 | - | 6 | 746 | |||
BC011579.1 | - | 6 | 729 | |||
BC090039.1 | - | 7 | 877 | 6181849 | ||
BX394857.2 | - | 7 | 780 |