Search
Search

SPSB2| Gene Symbol: SPSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146316.1 | NP_001139788.1 | 2 | 348 | ||
| NM_001319670.1 | NP_001306599.1 | 1 | 648 | |||
| NM_032641.3 | NP_116030.1 | 2 | 400 | |||
| GenBank mRNA | AF403027.1 | - | 1 | 192 | ||
| AK297963.1 | - | 2 | 343 | |||
| AK310353.1 | - | 1 | 597 | |||
| BC002983.1 | - | 2 | 331 | 3546861 | ||
| BC071933.1 | - | 2 | 348 | 6302511 | ||