Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300854.1 | NP_001287783.1 | 6 | 602 | ||
NM_053283.3 | NP_444513.1 | 5 | 538 | |||
GenBank mRNA | AF144011.1 | - | 5 | 408 | ||
AY590150.1 | - | 5 | 402 | |||
BC062682.1 | - | 5 | 410 | 30369929 | ||
BC069108.1 | - | 5 | 408 | 7216925 | ||
BP313072.1 | - | 5 | 410 | |||
EF503687.1 | - | 5 | 538 | |||
EF503688.1 | - | 6 | 705 | |||
EF503689.1 | - | 6 | 602 |