Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GCOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX095570.1 | - | 1 | 70 |
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018100.4 | NP_001018110.1 | 13 | 1866 | ||
NM_152451.7 | NP_689664.3 | 12 | 1782 | |||
GenBank mRNA | AK055535.1 | - | 13 | 1829 | ||
AL833028.1 | - | 8 | 1109 | |||
AL833142.1 | - | 11 | 1563 | |||
FJ970029.1 | - | 13 | 1743 |