Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMK2G SIRNA Assays ›
Gene Symbol:
CAMK2G
Gene Name:
calcium/calmodulin dependent protein kinase II gamma
Gene Aliases:
CAMK, CAMK-II, CAMKG
Chromosome Location:
Chr.10: 73812501 - 73874591 on Build GRCh38
UniGene ID:
Hs.523045
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
40719
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMK2G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204492.1
NP_001191421.1
18
1365
NM_001222.3
NP_001213.2
16
1233
NM_001320898.1
NP_001307827.1
19
1443
NM_172169.2
NP_751909.1
18
1329
NM_172170.4
NP_751910.1
17
1302
NM_172171.2
NP_751911.1
18
1416
NM_172173.2
NP_751913.1
17
1260
XM_005270195.1
XP_005270252.1
19
1470
XM_005270197.1
XP_005270254.1
19
1434
XM_005270198.1
XP_005270255.1
19
1428
XM_005270199.1
XP_005270256.1
18
1401
XM_005270200.1
XP_005270257.1
18
1371
XM_005270201.1
XP_005270258.1
18
1365
XM_005270203.1
XP_005270260.1
18
1326
XM_005270205.1
XP_005270262.1
17
1287
XM_006717993.1
XP_006718056.1
20
1503
XM_006717996.1
XP_006718059.1
19
1440
XM_006717997.1
XP_006718060.1
19
1389
XM_006717998.1
XP_006718061.1
19
1347
XM_011540225.2
XP_011538527.1
18
1355
XM_017016716.1
XP_016872205.1
20
1503
XM_017016717.1
XP_016872206.1
20
1451
XM_017016718.1
XP_016872207.1
19
1470
XM_017016719.1
XP_016872208.1
20
1461
XM_017016720.1
XP_016872209.1
19
1440
XM_017016721.1
XP_016872210.1
20
1578
XM_017016722.1
XP_016872211.1
19
1428
XM_017016723.1
XP_016872212.1
18
1407
XM_017016724.1
XP_016872213.1
18
1401
XM_017016725.1
XP_016872214.1
18
1371
XM_017016726.1
XP_016872215.1
18
1365
XM_017016727.1
XP_016872216.1
17
1338
XM_017016728.1
XP_016872217.1
18
1355
XM_017016729.1
XP_016872218.1
18
1238
XM_017016730.1
XP_016872219.1
18
1284
XM_017016731.1
XP_016872220.1
18
1346
XM_017016732.1
XP_016872221.1
18
1276
XM_017016733.1
XP_016872222.1
17
1241
XM_017016734.1
XP_016872223.1
18
1366
XM_017016735.1
XP_016872224.1
17
1257
XM_017016736.1
XP_016872225.1
18
1395
XM_017016737.1
XP_016872226.1
17
1309
XM_017016738.1
XP_016872227.1
19
1369
XM_017016739.1
XP_016872228.1
20
1635
GenBank mRNA
AB209474.1
-
3
5342
AK094774.1
-
3
1872
AK095576.1
-
19
2142
AK131199.1
-
18
1339
AK225771.1
-
18
1365
AK291704.1
-
17
1294
BC021269.2
-
15
977
BC034044.1
-
18
1320
4813948
L07043.1
-
14
908
L07044.1
-
17
1193
U50359.1
-
7
314
U50360.1
-
6
227
U66063.1
-
8
461
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
MAPK cascade
protein dephosphorylation
calcium ion transport
nervous system development
regulation of skeletal muscle adaptation
insulin secretion
cell differentiation
positive regulation of GTPase activity
protein autophosphorylation
protein oligomerization
regulation of calcium ion transport
interferon-gamma-mediated signaling pathway
regulation of cellular response to heat
regulation of relaxation of cardiac muscle
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
calcium-dependent protein serine/threonine phosphatase activity
Ras guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-4b6rj:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0