Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ASPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018136.4 | NP_060606.3 | 18 | 7195 | ||
GenBank mRNA | AF509326.1 | - | 18 | 6938 | ||
AK001379.1 | - | 1 | 324 | |||
AK095892.1 | - | 1 | 1983 | |||
AY099890.1 | - | 1 | 2589 | |||
AY367065.1 | - | 18 | 6938 | |||
AY971957.1 | - | 4 | 896 | |||
BX648804.1 | - | 1 | 2049 |