Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OXNAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138381.3 | NP_612390.1 | 5 | 777 | ||
XM_005265559.3 | XP_005265616.1 | 5 | 758 | |||
XM_006713405.1 | XP_006713468.1 | 5 | 741 | |||
XM_006713406.2 | XP_006713469.1 | 6 | 903 | |||
XM_011534230.2 | XP_011532532.1 | 5 | 758 | |||
XM_011534232.2 | XP_011532534.1 | 5 | 758 | |||
XM_011534233.2 | XP_011532535.1 | 5 | 758 | |||
XM_011534234.2 | XP_011532536.1 | 5 | 758 | |||
XM_011534235.2 | XP_011532537.1 | 5 | 758 | |||
XM_017007485.1 | XP_016862974.1 | 5 | 741 | |||
XM_017007486.1 | XP_016862975.1 | 5 | 773 | |||
XM_017007487.1 | XP_016862976.1 | 5 | 773 | |||
XM_017007488.1 | XP_016862977.1 | 5 | 773 | |||
XM_017007489.1 | XP_016862978.1 | 5 | 773 | |||
XM_017007490.1 | XP_016862979.1 | 5 | 773 | |||
XM_017007491.1 | XP_016862980.1 | 5 | 773 | |||
XM_017007492.1 | XP_016862981.1 | 5 | 512 | |||
XM_017007493.1 | XP_016862982.1 | 6 | 903 | |||
XM_017007494.1 | XP_016862983.1 | 6 | 903 | |||
XM_017007495.1 | XP_016862984.1 | 6 | 900 | |||
XM_017007496.1 | XP_016862985.1 | 6 | 900 | |||
GenBank mRNA | AB209556.1 | - | 3 | 271 | ||
AK298829.1 | - | 4 | 332 | |||
AK307258.1 | - | 4 | 640 | |||
AL583453.3 | - | 5 | 736 | |||
AL832787.1 | - | 1 | 79 | |||
BC008322.1 | - | 5 | 704 | 3358042 |