Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATXN2L SIRNA Assays ›
          Gene Symbol:
          ATXN2L
          Gene Name:
          ataxin 2 like
          Gene Aliases:
          A2D, A2LG, A2LP, A2RP
          Chromosome Location:
          Chr.16: 28823048 - 28837237 on Build GRCh38
          UniGene ID:
          Hs.460499
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 44518

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATXN2L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001308230.1 NP_001295159.1 2, 3 550
          NM_007245.3 NP_009176.2 2, 3 550
          NM_017492.3 NP_059867.3 2, 3 550
          NM_145714.2 NP_663760.1 2, 3 550
          NM_148414.2 NP_680780.1 2, 3 550
          NM_148415.2 NP_680781.1 2, 3 550
          NM_148416.2 NP_680782.1 2, 3 550
          XM_005255061.1 XP_005255118.1 2, 3 550
          XM_005255062.1 XP_005255119.1 2, 3 550
          XM_005255063.3 XP_005255120.1 2, 3 550
          XM_005255064.1 XP_005255121.1 2, 3 550
          XM_005255065.1 XP_005255122.1 2, 3 550
          XM_005255066.1 XP_005255123.1 2, 3 550
          XM_005255067.1 XP_005255124.1 2, 3 550
          XM_005255068.1 XP_005255125.1 2, 3 550
          XM_005255069.1 XP_005255126.1 2, 3 550
          XM_005255070.1 XP_005255127.1 2, 3 550
          XM_005255071.1 XP_005255128.1 2, 3 550
          XM_005255074.1 XP_005255131.1 2, 3 550
          XM_005255075.1 XP_005255132.1 2, 3 550
          XM_005255076.1 XP_005255133.1 2, 3 550
          XM_005255077.1 XP_005255134.1 2, 3 433
          XM_006721007.1 XP_006721070.1 2, 3 550
          XM_006721008.1 XP_006721071.1 2, 3 550
          XM_006721009.1 XP_006721072.1 2, 3 550
          XM_006721010.2 XP_006721073.1 2, 3 550
          XM_006721011.1 XP_006721074.1 2, 3 550
          XM_006721012.3 XP_006721075.1 2, 3 550
          XM_011545719.2 XP_011544021.1 2, 3 550
          XM_011545721.2 XP_011544023.1 2, 3 550
          XM_011545722.1 XP_011544024.1 2, 3 250
          XM_017022891.1 XP_016878380.1 2, 3 433
          XM_017022892.1 XP_016878381.1 2, 3 250
          GenBank mRNA AF034373.1 - 4, 5 652
          AJ317970.1 - 2, 3 506
          AJ317971.2 - 2, 3 506
          AJ317972.2 - 2, 3 506
          AJ317973.2 - 2, 3 506
          AJ317974.2 - 2, 3 506
          AK289981.1 - 2, 3 550
          AY188334.1 - 4, 5 652
          AY188335.1 - 4, 5 652
          AY188336.1 - 4, 5 652
          AY188337.1 - 4, 5 652
          AY188338.1 - 4, 5 652
          BC068012.1 - 3, 4 408
          BC082760.1 - 2, 3 346 5812230
          BC136584.1 - 2, 3 503 9020574
          U70671.1 - 2, 3 180

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cytoplasmic mRNA processing body assembly
          stress granule assembly
          cell-cell adhesion
          protein binding
          poly(A) RNA binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0