Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127711.2 | NP_001121183.1 | 4 | 315 | ||
NM_001286484.1 | NP_001273413.1 | 4 | 340 | |||
NM_001286488.1 | NP_001273417.1 | 5 | 417 | |||
NM_001286489.1 | NP_001273418.1 | 4 | 315 | |||
NM_030969.4 | NP_112231.3 | 5 | 417 | |||
NR_104454.1 | 3 | 238 | ||||
GenBank mRNA | AK294152.1 | - | 5 | 371 | ||
BC001033.1 | - | 5 | 359 | 3139565 | ||
BC013913.1 | - | 5 | 379 | 3685635 | ||
BC071660.1 | - | 5 | 361 | 4473732 | ||
BG288654.1 | - | 4 | 268 | |||
BG432075.1 | - | 5 | 346 | |||
BP249229.1 | - | 4 | 269 | |||
BQ056988.1 | - | 4 | 268 | |||
BU954633.1 | - | 3 | 180 |