Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MIF SIRNA Assays ›
          Gene Symbol:
          Mif LOC103694877
          Gene Name:
          macrophage migration inhibitory factor (glycosylation-inhibiting factor)
          macrophage migration inhibitory factor
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.20: 13732114 - 13732979 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906821: 11287 - 12316 on Build Rnor_6.0
          UniGene ID:
          Rn.2661
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 46855

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mif
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031051.1 NP_112313.1 2 229
          GenBank mRNA BC061545.1 - 2 213 6920605
          FQ210474.1 - 2 211
          FQ213295.1 - 2 198
          FQ217414.1 - 2 209
          FQ218686.1 - 2 198
          FQ222835.1 - 2 225
          FQ223656.1 - 2 186
          FQ229019.1 - 2 209
          FQ229084.1 - 2 209
          S73424.1 - 2 194
          U62326.1 - 2 229
          Gene Symbol: LOC103694877
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008773846.1 XP_008772068.1 2 393

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          decarboxylase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          positive regulation of protein phosphorylation
          hair follicle development
          brain renin-angiotensin system
          positive regulation of acute inflammatory response
          negative regulation of mature B cell apoptotic process
          inflammatory response
          cell surface receptor signaling pathway
          spermatogenesis
          brain development
          aging
          cell aging
          metabolic process
          cell proliferation
          response to heat
          response to mechanical stimulus
          response to hormone
          response to inorganic substance
          negative regulation of gene expression
          positive regulation of protein kinase A signaling
          positive regulation of smooth muscle cell migration
          carboxylic acid metabolic process
          DNA damage response, signal transduction by p53 class mediator
          positive regulation of B cell proliferation
          positive regulation of lipopolysaccharide-mediated signaling pathway
          negative regulation of cellular protein metabolic process
          response to estradiol
          response to lipopolysaccharide
          response to progesterone
          response to menaquinone
          response to insulin
          negative regulation of myeloid cell apoptotic process
          positive regulation of peptidyl-serine phosphorylation
          response to vitamin E
          wound healing
          regulation of cell proliferation
          positive regulation of phosphorylation
          response to drug
          drinking behavior
          negative regulation of apoptotic process
          positive regulation of potassium ion transport
          positive regulation of MAP kinase activity
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          skin development
          innate immune response
          positive regulation of glycolytic process
          positive regulation of glucose import
          positive regulation of fibroblast proliferation
          positive regulation of axon regeneration
          positive regulation of cytokine secretion
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of immune response
          positive chemotaxis
          response to glucocorticoid
          positive regulation of prostaglandin secretion involved in immune response
          positive regulation of myeloid leukocyte cytokine production involved in immune response
          protein homotrimerization
          cellular response to hydrogen peroxide
          positive regulation of ERK1 and ERK2 cascade
          negative regulation of cell cycle arrest
          cellular response to hypoxia
          positive regulation of arachidonic acid secretion
          negative regulation of cell aging
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of chemokine (C-X-C motif) ligand 2 production
          dopachrome isomerase activity
          receptor binding
          cytokine activity
          cytokine receptor binding
          protein complex binding
          chemoattractant activity
          phenylpyruvate tautomerase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0