Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: D17Wsu92e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033279.3 | NP_001028451.2 | 4 | 1023 | ||
GenBank mRNA | AK143016.1 | - | 4 | 935 | ||
AK161603.1 | - | 4 | 1024 | |||
AK165011.1 | - | 4 | 1023 | |||
BC132324.1 | - | 5 | 949 | 40130601 | ||
BC132326.1 | - | 5 | 949 | 40130603 | ||
BC145403.1 | - | 4 | 1023 | 9053961 |