Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: L3mbtl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081338.1 | NP_001074807.1 | 23 | 2657 | ||
GenBank mRNA | AK220350.1 | - | 21 | 2405 | ||
BC116639.1 | - | 20 | 3071 | 40061282 |