Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kif26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097621.1 | NP_001091090.1 | 16 | 6886 | ||
GenBank mRNA | AK129315.1 | - | 8 | 5011 | ||
BC059062.1 | - | 5 | 3718 |