Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DLG4 SIRNA Assays ›
          Gene Symbol:
          Dlg4
          Gene Name:
          discs, large homolog 4 (Drosophila)
          Gene Aliases:
          Dlgh4, PSD95, Sap90
          Chromosome Location:
          Chr.10: 56625630 - 56655543 on Build Rnor_6.0
          UniGene ID:
          Rn.9765
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 51054

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dlg4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019621.1 NP_062567.1 5, 6 400
          XM_006246680.2 XP_006246742.1 7, 8 776
          XM_006246681.2 XP_006246743.1 7, 8 771
          XM_006246682.2 XP_006246744.1 7, 8 625
          XM_006246683.2 XP_006246745.1 7, 8 533
          XM_006246684.2 XP_006246746.1 7, 8 524
          XM_006246685.2 XP_006246747.1 5, 6 486
          XM_006246686.2 XP_006246748.1 5, 6 546
          XM_006246687.2 XP_006246749.1 5, 6 446
          XM_006246688.2 XP_006246750.1 4, 5 2647
          XM_006246689.2 XP_006246751.1 4, 5 389
          XM_006246691.2 XP_006246753.1 4, 5 380
          XM_008767809.1 XP_008766031.1 7, 8 877
          XM_008767810.1 XP_008766032.1 4, 5 264
          GenBank mRNA M96853.1 - 5, 6 400
          X66474.1 - 5, 6 545

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of receptor internalization
          protein complex assembly
          positive regulation of cytosolic calcium ion concentration
          synaptic vesicle maturation
          social behavior
          protein localization to synapse
          locomotory exploration behavior
          neuronal ion channel clustering
          establishment of protein localization
          establishment or maintenance of epithelial cell apical/basal polarity
          nucleotide phosphorylation
          regulation of long-term neuronal synaptic plasticity
          positive regulation of synaptic transmission
          neuromuscular process controlling balance
          dendritic spine morphogenesis
          vocalization behavior
          AMPA glutamate receptor clustering
          receptor localization to synapse
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          positive regulation of excitatory postsynaptic potential
          regulation of grooming behavior
          guanylate kinase activity
          receptor binding
          protein binding
          protein C-terminus binding
          protein kinase binding
          protein phosphatase binding
          PDZ domain binding
          beta-1 adrenergic receptor binding
          D1 dopamine receptor binding
          P2Y1 nucleotide receptor binding
          protein complex binding
          acetylcholine receptor binding
          glutamate receptor binding
          ionotropic glutamate receptor binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline