Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DBI SIRNA Assays ›
          Gene Symbol:
          Dbi
          Gene Name:
          diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein)
          Gene Aliases:
          Acbp, Acoabp3, Ep, Odn, RNACOABP3, Ttn
          Chromosome Location:
          Chr.13: 36147667 - 36156076 on Build Rnor_6.0
          UniGene ID:
          Rn.3285
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 54498

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dbi
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031853.4 NP_114054.1 4 326
          GenBank mRNA AY383701.1 - 506
          BC084717.1 - 3 213 7310272
          FQ209835.1 - 4 274
          FQ209938.1 - 4 269
          FQ210140.1 - 4 274
          FQ210336.1 - 4 271
          FQ210460.1 - 4 274
          FQ210781.1 - 4 301
          FQ210997.1 - 4 271
          FQ211081.1 - 4 274
          FQ211192.1 - 4 274
          FQ211339.1 - 4 275
          FQ211377.1 - 3 195
          FQ213527.1 - 4 275
          FQ214572.1 - 4 274
          FQ217714.1 - 4 275
          FQ218148.1 - 4 269
          FQ218164.1 - 4 293
          FQ218239.1 - 4 274
          FQ218300.1 - 4 274
          FQ218680.1 - 4 274
          FQ219136.1 - 4 274
          FQ219739.1 - 4 274
          FQ221153.1 - 4 269
          FQ221871.1 - 4 275
          FQ222052.1 - 4 274
          FQ222378.1 - 4 276
          FQ222416.1 - 4 269
          FQ222549.1 - 4 264
          FQ222564.1 - 4 275
          FQ222579.1 - 4 305
          FQ223167.1 - 4 269
          FQ223413.1 - 4 274
          FQ223492.1 - 4 275
          FQ223501.1 - 4 274
          FQ223602.1 - 4 232
          FQ224361.1 - 4 274
          FQ224797.1 - 4 275
          FQ229114.1 - 4 269
          M14201.1 - 4 326
          M20268.1 - 4 215

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          hair follicle development
          acyl-CoA metabolic process
          triglyceride metabolic process
          steroid biosynthetic process
          brain development
          learning or memory
          glial cell proliferation
          protein palmitoylation
          lateral ventricle development
          pancreatic juice secretion
          cellular response to nutrient
          negative regulation of fatty acid beta-oxidation
          regulation of synaptic transmission, GABAergic
          phosphatidylcholine acyl-chain remodeling
          skin development
          positive regulation of lipid biosynthetic process
          positive regulation of release of sequestered calcium ion into cytosol
          long-term synaptic potentiation
          fatty-acyl-CoA binding
          protein binding
          lipid binding
          benzodiazepine receptor binding
          long-chain fatty acyl-CoA binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0