Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GRN SIRNA Assays ›
          Gene Symbol:
          Grn
          Gene Name:
          granulin
          Gene Aliases:
          Pgrn, epithelin
          Chromosome Location:
          Chr.11: 102430322 - 102436809 on Build GRCm38
          UniGene ID:
          Mm.1568
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 61978

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Grn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008175.4 NP_032201.2 6, 7 824
          GenBank mRNA AK013678.1 - 6, 7 640
          AK018744.1 - 6, 7 665
          AK147932.1 - 6, 7 641
          AK149570.1 - 6, 7 643
          AK150125.1 - 6, 7 639
          AK150205.1 - 6, 7 640
          AK150338.1 - 6, 7 629
          AK150420.1 - 6, 7 628
          AK150717.1 - 6, 7 640
          AK150746.1 - 6, 7 641
          AK150881.1 - 6, 7 641
          AK151007.1 - 6, 7 640
          AK151162.1 - 6, 7 641
          AK151343.1 - 6, 7 640
          AK151713.1 - 6, 7 640
          AK151756.1 - 6, 7 640
          AK152048.1 - 6, 7 640
          AK152087.1 - 6, 7 640
          AK152118.1 - 6, 7 629
          AK152213.1 - 6, 7 629
          AK152705.1 - 6, 7 640
          AK152765.1 - 6, 7 640
          AK152944.1 - 6, 7 629
          AK153183.1 - 6, 7 629
          AK153269.1 - 6, 7 640
          AK153289.1 - 6, 7 640
          AK153418.1 - 6, 7 640
          AK153470.1 - 6, 7 640
          AK154492.1 - 6, 7 640
          AK159397.1 - 6, 7 629
          AK159428.1 - 6, 7 629
          AK159557.1 - 6, 7 640
          AK159809.1 - 6, 7 640
          AK160022.1 - 6, 7 629
          AK160033.1 - 6, 7 640
          AK160074.1 - 6, 7 629
          AK211032.1 - 1, 2 92
          AK219696.1 - 1, 2 103
          BC037047.1 - 5, 6 621
          BC049943.1 - 6, 7 626
          BC129849.1 - 6, 7 624 40109630
          BC129850.1 - 6, 7 624 40109631
          M86736.1 - 6, 7 622
          X62321.1 - 5, 6 605

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst hatching
          embryo implantation
          positive regulation of neuron projection development
          response to estradiol
          chondrocyte proliferation
          synaptic vesicle endocytosis
          positive regulation of epithelial cell proliferation
          male mating behavior
          positive regulation of dendritic spine development
          neural precursor cell proliferation
          cytokine activity
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-2fl6k:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline