Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008313.4 | NP_032339.2 | 6 | 738 | ||
GenBank mRNA | AJ011369.1 | - | 5 | 528 | ||
AK141587.1 | - | 6 | 682 | |||
AK141625.1 | - | 6 | 739 | |||
BC132189.1 | - | 5 | 567 | |||
BC132191.1 | - | 5 | 567 | |||
BC145159.1 | - | 5 | 567 | |||
BC145161.1 | - | 5 | 567 | |||
Y09585.1 | - | 5 | 528 | |||
Y09587.1 | - | 5 | 528 | |||
Y09588.1 | - | 5 | 528 |
Set Membership: |
G-protein coupled receptor |