Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRP6 SIRNA Assays ›
          Gene Symbol:
          Lrp6
          Gene Name:
          low density lipoprotein receptor-related protein 6
          Gene Aliases:
          C030016K15Rik, Cd, Gw, ska26, ska<m26Jus>, skax26
          Chromosome Location:
          Chr.6: 134446476 - 134566960 on Build GRCm38
          UniGene ID:
          Mm.321990
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 62887

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Lrp6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008514.4 NP_032540.2 15 3455
          GenBank mRNA AF074265.1 - 15 3324
          AK140796.1 - 3 850
          AK162203.1 - 2 259
          BC060704.1 - 15 3455 6409058

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          gastrulation with mouth forming second
          somitogenesis
          neural tube closure
          negative regulation of protein phosphorylation
          heart looping
          morphogenesis of an epithelium
          positive regulation of mesenchymal cell proliferation
          pericardium morphogenesis
          axis elongation
          regulation of transcription, DNA-templated
          negative regulation of protein kinase activity
          endocytosis
          receptor-mediated endocytosis
          positive regulation of cytosolic calcium ion concentration
          chemical synaptic transmission
          multicellular organism development
          embryonic pattern specification
          dorsal/ventral axis specification
          anterior/posterior pattern specification
          neural crest formation
          neural crest cell differentiation
          Wnt signaling pathway
          single organismal cell-cell adhesion
          cerebellum morphogenesis
          thalamus development
          cerebral cortex cell migration
          forebrain radial glial cell differentiation
          forebrain generation of neurons
          Wnt signaling pathway involved in forebrain neuroblast division
          neural tube development
          formation of radial glial scaffolds
          cerebral cortex development
          negative regulation of Wnt signaling pathway
          embryonic limb morphogenesis
          forebrain development
          midbrain development
          midbrain-hindbrain boundary development
          negative regulation of smooth muscle cell apoptotic process
          limb morphogenesis
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          external genitalia morphogenesis
          post-anal tail morphogenesis
          cell migration involved in gastrulation
          regulation of cell proliferation
          odontogenesis of dentin-containing tooth
          embryonic digit morphogenesis
          positive regulation of apoptotic process
          Wnt signaling pathway involved in dorsal/ventral axis specification
          negative regulation of fat cell differentiation
          positive regulation of ossification
          positive regulation of bone resorption
          positive regulation of cell cycle
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          bone remodeling
          embryonic camera-type eye morphogenesis
          generation of neurons
          skeletal system morphogenesis
          negative regulation of epithelial cell proliferation
          positive regulation of sequence-specific DNA binding transcription factor activity
          response to folic acid
          palate development
          convergent extension
          retina morphogenesis in camera-type eye
          embryonic retina morphogenesis in camera-type eye
          canonical Wnt signaling pathway
          regulation of cell development
          face morphogenesis
          bone morphogenesis
          branching involved in mammary gland duct morphogenesis
          trachea cartilage morphogenesis
          mammary placode formation
          mammary gland duct morphogenesis
          canonical Wnt signaling pathway involved in cardiac neural crest cell differentiation involved in heart development
          canonical Wnt signaling pathway involved in positive regulation of cardiac outflow tract cell proliferation
          cellular response to cholesterol
          dopaminergic neuron differentiation
          negative regulation of protein serine/threonine kinase activity
          primitive streak formation
          receptor-mediated endocytosis of low-density lipoprotein particle involved in cholesterol transport
          Wnt signaling pathway involved in somitogenesis
          axis elongation involved in somitogenesis
          positive regulation of canonical Wnt signaling pathway
          toxin transport
          negative regulation of non-canonical Wnt signaling pathway
          positive regulation of Wnt signaling pathway involved in dorsal/ventral axis specification
          negative regulation of planar cell polarity pathway involved in ventricular septum morphogenesis
          negative regulation of planar cell polarity pathway involved in cardiac muscle tissue morphogenesis
          negative regulation of planar cell polarity pathway involved in cardiac right atrium morphogenesis
          negative regulation of planar cell polarity pathway involved in outflow tract morphogenesis
          negative regulation of planar cell polarity pathway involved in pericardium morphogenesis
          negative regulation of planar cell polarity pathway involved in neural tube closure
          low-density lipoprotein receptor activity
          receptor binding
          frizzled binding
          protein binding
          Wnt-protein binding
          kinase inhibitor activity
          toxin transporter activity
          apolipoprotein binding
          identical protein binding
          protein homodimerization activity
          Wnt-activated receptor activity
          coreceptor activity involved in Wnt signaling pathway

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0