Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FOXH1 SIRNA Assays ›
Gene Symbol:
FOXH1
Gene Name:
forkhead box H1
Gene Aliases:
FAST-1, FAST1
Chromosome Location:
Chr.8: 144473732 - 144476335 on Build GRCh38
UniGene ID:
Hs.708365
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
6426
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FOXH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003923.2
NP_003914.1
1
655
GenBank mRNA
AF076292.1
-
1
655
BC065377.1
-
1
158
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
heart looping
secondary heart field specification
outflow tract morphogenesis
cardiac right ventricle morphogenesis
ventricular trabecula myocardium morphogenesis
transcription from RNA polymerase II promoter
transforming growth factor beta receptor signaling pathway
negative regulation of intracellular estrogen receptor signaling pathway
embryonic heart tube anterior/posterior pattern specification
aorta morphogenesis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
axial mesoderm development
negative regulation of androgen receptor signaling pathway
nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
negative regulation of androgen receptor activity
RNA polymerase II transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription cofactor activity
transcriptional activator activity, RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein domain specific binding
enhancer binding
bHLH transcription factor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
SMAD binding
androgen receptor binding
co-SMAD binding
R-SMAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0