Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANGPT1 SIRNA Assays ›
          Gene Symbol:
          Angpt1
          Gene Name:
          angiopoietin 1
          Gene Aliases:
          1110046O21Rik, Ang-1, Ang1
          Chromosome Location:
          Chr.15: 42424667 - 42676977 on Build GRCm38
          UniGene ID:
          Mm.309336
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 65670

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Angpt1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286062.1 NP_001272991.1 4 1130
          NM_009640.4 NP_033770.2 4 1130
          GenBank mRNA AK012027.1 - 4 1066
          AK088439.1 - 4 1131
          AK172868.1 - 4 950
          BC067410.1 - 4 948 30436666
          U83509.1 - 4 920

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          ovarian follicle development
          patterning of blood vessels
          vasculogenesis
          in utero embryonic development
          negative regulation of protein phosphorylation
          sprouting angiogenesis
          positive regulation of receptor internalization
          negative regulation of cytokine secretion involved in immune response
          endocardium morphogenesis
          negative regulation of cell adhesion
          transmembrane receptor protein tyrosine kinase signaling pathway
          activation of transmembrane receptor protein tyrosine kinase activity
          multicellular organism development
          endoderm development
          positive regulation of endothelial cell migration
          positive regulation of phosphatidylinositol 3-kinase signaling
          regulation of skeletal muscle satellite cell proliferation
          hemopoiesis
          cell differentiation
          heparin biosynthetic process
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          positive regulation of protein ubiquitination
          cell-substrate adhesion
          regulation of tumor necrosis factor production
          positive regulation of peptidyl-serine phosphorylation
          protein localization to cell surface
          negative regulation of protein import into nucleus
          negative regulation of apoptotic process
          negative regulation of vascular permeability
          regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of protein binding
          negative regulation of neuron apoptotic process
          positive regulation of blood vessel endothelial cell migration
          positive regulation of cell adhesion
          Tie signaling pathway
          positive regulation of peptidyl-tyrosine phosphorylation
          positive chemotaxis
          protein homooligomerization
          positive regulation of protein kinase B signaling
          cardiac muscle tissue morphogenesis
          vasculogenesis involved in coronary vascular morphogenesis
          positive regulation of ERK1 and ERK2 cascade
          glomerulus vasculature development
          negative regulation of endothelial cell apoptotic process
          regulation of macrophage migration inhibitory factor signaling pathway
          receptor binding
          vascular endothelial growth factor receptor binding
          receptor tyrosine kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-sg7z8:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline