Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CACNA1D SIRNA Assays ›
          Gene Symbol:
          CACNA1D
          Gene Name:
          calcium voltage-gated channel subunit alpha1 D
          Gene Aliases:
          CACH3, CACN4, CACNL1A2, CCHL1A2, Cav1.3, PASNA, SANDD
          Chromosome Location:
          Chr.3: 53495049 - 53813151 on Build GRCh38
          UniGene ID:
          Hs.476358
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 7032

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CACNA1D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000720.3 NP_000711.1 2 211
          NM_001128839.2 NP_001122311.1 2 211
          NM_001128840.2 NP_001122312.1 2 211
          XM_005265448.3 XP_005265505.1 2 93
          XM_011534094.2 XP_011532396.1 2 219
          XM_011534096.2 XP_011532398.1 2 219
          XM_011534100.2 XP_011532402.1 2 219
          XM_017007137.1 XP_016862626.1 2 219
          XM_017007138.1 XP_016862627.1 2 219
          XM_017007139.1 XP_016862628.1 2 219
          XM_017007140.1 XP_016862629.1 2 219
          XM_017007141.1 XP_016862630.1 2 219
          XM_017007142.1 XP_016862631.1 2 219
          XM_017007143.1 XP_016862632.1 2 219
          XM_017007144.1 XP_016862633.1 2 219
          XM_017007145.1 XP_016862634.1 2 219
          GenBank mRNA EU363339.1 - 2 93
          M76558.1 - 2 603
          M83566.1 - 2 211

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          sensory perception of sound
          regulation of insulin secretion
          positive regulation of calcium ion transport
          regulation of atrial cardiac muscle cell membrane repolarization
          cardiac conduction
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during cardiac muscle cell action potential
          membrane depolarization during SA node cell action potential
          regulation of heart rate by cardiac conduction
          regulation of potassium ion transmembrane transporter activity
          regulation of potassium ion transmembrane transport
          voltage-gated calcium channel activity
          high voltage-gated calcium channel activity
          ankyrin binding
          metal ion binding
          alpha-actinin binding
          voltage-gated calcium channel activity involved in cardiac muscle cell action potential
          voltage-gated calcium channel activity involved SA node cell action potential

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0