Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NT5C SIRNA Assays ›
          Gene Symbol:
          Nt5c
          Gene Name:
          5',3'-nucleotidase, cytosolic
          Gene Aliases:
          Dnt, Dnt1, Umph-2, Umph2
          Chromosome Location:
          Chr.11: 115490425 - 115491876 on Build GRCm38
          UniGene ID:
          Mm.390379
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 72317

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nt5c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015807.1 NP_056622.1 5 751
          GenBank mRNA AF078840.1 - 5 751
          AK007418.1 - 5 790
          BC024368.1 - 5 754 5065485

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          nucleotide metabolic process
          pyrimidine deoxyribonucleotide catabolic process
          deoxyribonucleotide catabolic process
          dephosphorylation
          nucleotide binding
          protein tyrosine/serine/threonine phosphatase activity
          nucleotidase activity
          5'-nucleotidase activity
          protein tyrosine/threonine phosphatase activity
          JUN kinase phosphatase activity
          phosphohistidine phosphatase activity
          inositol bisphosphate phosphatase activity
          hydrolase activity
          phosphatase activity
          MAP kinase tyrosine/serine/threonine phosphatase activity
          inositol-1,3,4-trisphosphate 4-phosphatase activity
          pyrimidine nucleotide binding
          NADP phosphatase activity
          transmembrane receptor protein phosphatase activity
          inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
          inositol-1,4,5,6-tetrakisphosphate 6-phosphatase activity
          inositol-4,5-bisphosphate 5-phosphatase activity
          5-amino-6-(5-phosphoribitylamino)uracil phosphatase activity
          phosphatidylinositol-4-phosphate phosphatase activity
          phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
          metal ion binding
          phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
          inositol phosphate phosphatase activity
          inositol-1,3,4,5,6-pentakisphosphate 1-phosphatase activity
          inositol-3,4-bisphosphate 4-phosphatase activity
          inositol-1,3,4-trisphosphate 1-phosphatase activity
          inositol-1,3,4,6-tetrakisphosphate 6-phosphatase activity
          inositol-1,3,4,6-tetrakisphosphate 1-phosphatase activity
          phosphatidylinositol-1,4,5-trisphosphate 5-phosphatase activity
          IDP phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8bdhq:80/100.66.78.62:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0