Search
Search

Emcn| Gene Symbol: Emcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163522.1 | NP_001156994.1 | 2, 3 | 366 | ||
| NM_016885.2 | NP_058581.2 | 2, 3 | 366 | |||
| GenBank mRNA | AA822842.1 | - | 2, 3 | 366 | ||
| AB034693.1 | - | 2, 3 | 319 | |||
| AF060883.1 | - | 2, 3 | 301 | |||
| AK002616.1 | - | 2, 3 | 348 | |||
| AK029348.1 | - | 334 | ||||
| AK145825.1 | - | 2, 3 | 355 | |||
| AK164422.1 | - | 287 | ||||
| BC003706.1 | - | 2, 3 | 327 | 3485788 | ||